3-(4-Fluorophenyl)oxetane-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
115898-250MG
115898-100MG
115898-500MG
102898-684EA
2843.08
USD
102898-684
102898-686
102898-682
3-(4-Fluorophenyl)oxetane-3-carboxylic acid ≥95%
3-(4-Fluorophenyl)oxetane-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₉FO₃ |
MDL Number:
MFCD22544051 CAS Number: 1393572-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...