3-Chloro-5-methoxypyridazine
Supplier: Matrix Scientific
113085-1G
113085-5G
102891-440EA
1200
USD
102891-440
102897-706
3-Chloro-5-methoxypyridazine
3-Chloro-5-methoxypyridazine
Hazard: Irritant
Formula:
C₅H₅ClN₂O MW: 144.5600 g/mol |
MDL Number:
MFCD08236839 CAS Number: 123696-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...