tert-Butyl-4-[methyl(tetrahydropyran-4-yl)amino]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115531-250MG 115531-500MG 115531-1G
102896-656EA 620 USD
102896-656 102896-658 102896-660
tert-Butyl-4-[methyl(tetrahydropyran-4-yl)amino]piperidine-1-carboxylate ≥95%
tert-Butyl-4-[methyl(tetrahydropyran-4-yl)amino]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₃₀N₂O₃
MW: 298.43 g/mol
MDL Number: MFCD22370062
CAS Number: 1232060-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR