2,4-Difluoro-3-(trifluoromethyl)bromobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-2,4-difluoro-3-(trifluoromethyl)benzene3-Bromo-2,6-difluorobenzotrifluoride

114044-1G
102893-200EA 813.85 USD
102893-200
2,4-Difluoro-3-(trifluoromethyl)bromobenzene ≥97%
2,4-Difluoro-3-(trifluoromethyl)bromobenzene

Hazard: Irritant

Formula: C₇H₂BrF₅
MW: 260.99 g/mol
MDL Number: MFCD18379808
CAS Number: 1263377-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR