Methyl-4-butylbenzoate ≥97%

Supplier: Matrix Scientific

113921-1G
102893-000EA 117.3 USD
102893-000
Methyl-4-butylbenzoate ≥97%
Methyl 4-butylbenzoate

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD00277209
CAS Number: 20651-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR