4,5-Difluorosalicylaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-4,5-difluorobenzaldehyde4,5-Difluoro-2-hydroxybenzaldehyde

113562-1G
102892-440EA 299 USD
102892-440
4,5-Difluorosalicylaldehyde ≥97%
4,5-Difluorosalicylaldehyde

Hazard: Irritant

Formula: C₇H₄F₂O₂
MW: 158.11 g/mol
MDL Number: MFCD08458005
CAS Number: 199287-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR