1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid

Supplier: Matrix Scientific

113054-1G
102897-600EA 1629.23 USD
102897-600
1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid
1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₅F₂NO₄
MDL Number: MFCD22415254
CAS Number: 1363380-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR