5-Bromo-2,7-dimethyl-2H-indazole ≥95%

Supplier: Matrix Scientific

116015-1G
102900-208EA 787.69 USD
102900-208
5-Bromo-2,7-dimethyl-2H-indazole ≥95%
5-Bromo-2,7-dimethyl-2H-indazole

Hazard: Irritant

Formula: C₉H₉BrN₂
MDL Number: MFCD12028637
CAS Number: 1146637-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR