1-((4-Azidobutoxy)methyl)-4-methoxybenzene ≥97%

Supplier: Matrix Scientific

116006-5G
102900-108EA 983.08 USD
102900-108
1-((4-Azidobutoxy)methyl)-4-methoxybenzene ≥97%
1-((4-Azidobutoxy)methyl)-4-methoxybenzene

Hazard: Irritant

Formula: C₁₂H₁₈N₃O₂
MDL Number: MFCD26792595
CAS Number: 1355510-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR