4-Azidobutyrate-N-hydroxysuccinimide ester ≥97%

Supplier: Matrix Scientific

115998-100MG
102900-362EA 569.23 USD
102900-362
4-Azidobutyrate-N-hydroxysuccinimide ester ≥97%
4-Azidobutyrate-N-hydroxysuccinimide ester

Hazard: Irritant

Formula: C₈H₁₀N₄O₄
MDL Number: MFCD22573606
CAS Number: 943858-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR