4-Carboxypennsylvania green methyl ester ≥96%

Supplier: Matrix Scientific

115994-500MG 115994-100MG
102900-338EA 769.23 USD
102900-338 102900-342
4-Carboxypennsylvania green methyl ester ≥96%
4-Carboxypennsylvania green methyl ester

Hazard: Irritant

Formula: C₂₂H₁₄F₂O₅
MW: 396.35 g/mol
MDL Number: MFCD26406706
CAS Number: 879288-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR