1,1-Dichloro-2-ethoxycyclopropane ≥95%

Supplier: Matrix Scientific

115993-1G
102900-334EA 330.77 USD
102900-334
1,1-Dichloro-2-ethoxycyclopropane ≥95%
1,1-Dichloro-2-ethoxycyclopropane

Hazard: Irritant

Formula: C₅H₈Cl₂O
MDL Number: MFCD01727254
CAS Number: 7363-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR