2-Chloro-3,3-diethoxyprop-1-ene ≥95%

Supplier: Matrix Scientific

115992-1G
102900-330EA 604.62 USD
102900-330
2-Chloro-3,3-diethoxyprop-1-ene ≥95%
2-Chloro-3,3-diethoxyprop-1-ene

Hazard: Irritant

Formula: C₇H₁₃ClO₂
MDL Number: MFCD23102091
CAS Number: 7575-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR