Coumarin boronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: B-(2-oxo-2H-1-benzopyran-7-yl)-boronic acidCBA(2-Oxochromen-7-yl)boronic acid

115985-250MG
102900-148EA 466.15 USD
102900-148
Coumarin boronic acid ≥96%
Coumarin boronic acid
Formula: C₉H₇BO₄
MW: 189.96 g/mol
MDL Number: MFCD25562900
CAS Number: 1357078-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR