2-Oxo-2H-chromen-7-yltrifluormethanesulfonate ≥97%

Supplier: Matrix Scientific

Synonyms: Trifluoromethanesulfonic acid 2-Oxo-2H-chromen-7-yl ester2-Oxo-2H-chromen-7-yl trifluoromethanesulphonate2-Oxo-2H-chromen-7-yl trifluoromethanesulfonate7-Coumaryl trifluoromethanesulphonate7-Coumaryl trifluoromethanesulfonate7-Coumaryl triflateTrifluoromethanesulphonic acid 2-Oxo-2H-chromen-7-yl ester

115981-5G
102900-124EA 1241.54 USD
102900-124
2-Oxo-2H-chromen-7-yltrifluormethanesulfonate ≥97%
2-Oxo-2H-chromen-7-yltrifluormethanesulfonate

Hazard: Irritant

Formula: C₁₀H₅F₃O₅S
MW: 294.21 g/mol
MDL Number: MFCD18382236
CAS Number: 108530-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR