5-(Tributylstannyl)-2-(trimethylsilyl)thiazole ≥95%

Supplier: Matrix Scientific

115976-500MG 115976-5G
102900-022EA 933.85 USD
102900-022 102900-026
5-(Tributylstannyl)-2-(trimethylsilyl)thiazole ≥95%
5-(Tributylstannyl)-2-(trimethylsilyl)thiazole

Hazard: Irritant

Formula: C₇H₁₃NSSi
MDL Number: MFCD16170327
CAS Number: 263868-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR