1-(2-Bromo-4-chlorophenyl)cyclopropylamine ≥95%

Supplier: Matrix Scientific

115971-1G
102900-060EA 1867.69 USD
102900-060
1-(2-Bromo-4-chlorophenyl)cyclopropylamine ≥95%
1-(2-Bromo-4-chlorophenyl)cyclopropylamine

Hazard: Irritant

Formula: C₉H₉BrClN
MDL Number: MFCD14525461
CAS Number: 1204580-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR