Tributylstannyliodomethane 90-95%

Supplier: Matrix Scientific

115970-1G
102900-054EA 150.77 USD
102900-054
Tributylstannyliodomethane 90-95%
Tributylstannyliodomethane

Hazard: Irritant

Formula: C₁₃H₂₉ISn
MDL Number: MFCD00161398
CAS Number: 66222-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR