2-Bromo-3-phenylpropionic acid ≥90%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3-phenylpropanoic acid

115965-1G
102899-930EA 84.62 USD
102899-930
2-Bromo-3-phenylpropionic acid ≥90%
2-Bromo-3-phenylpropionic acid

Hazard: Irritant/Lachrymator

Formula: C₉H₉BrO₂
MW: 229.08 g/mol
MDL Number: MFCD00461796
CAS Number: 16503-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR