8-Boc-2-fluoro-8-azabicyclo[3.2.1]octan-3-ol ≥95%

Supplier: Matrix Scientific

115960-1G
102899-986EA 1292.31 USD
102899-986
8-Boc-2-fluoro-8-azabicyclo[3.2.1]octan-3-ol ≥95%
8-Boc-2-fluoro-8-azabicyclo[3.2.1]octan-3-ol

Hazard: Irritant

Formula: C₁₂H₂₀FNO₃
MW: 245.3 g/mol
MDL Number: MFCD22580393
CAS Number: 1404196-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR