3-[3-(Trifluoromethyl)phenyl]oxetan-3-amine hydrochloride ≥95%
Supplier: Matrix Scientific
115956-5G
115956-1G
115956-500MG
102899-628EA
5767.69
USD
102899-628
102899-626
102899-624
3-[3-(Trifluoromethyl)phenyl]oxetan-3-amine hydrochloride ≥95%
3-[3-(Trifluoromethyl)phenyl]oxetan-3-amine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₁ClF₃NO |
MDL Number:
MFCD20233043 CAS Number: 1349719-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...