N-Trideuteromethyloxetan-3-amine ≥95%

Supplier: Matrix Scientific

115954-5G 115954-1G
102899-616EA 6907.69 USD
102899-616 102899-614
N-Trideuteromethyloxetan-3-amine ≥95%
N-Trideuteromethyloxetan-3-amine

Hazard: Irritant

Formula: C₄H₆D₃NO
CAS Number: 1403767-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR