1-(Oxetan-3-yl)-1H-pyrazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
115952-500MG
102900-244EA
1887.69
USD
102900-244
1-(Oxetan-3-yl)-1H-pyrazole-4-carboxylic acid ≥95%
1-(Oxetan-3-yl)-1H-pyrazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₇H₈N₂O₃ |
MDL Number:
MFCD21848617 CAS Number: 1389323-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...