(3S)-1-(Oxetan-3-yl)piperidin-3-amine ≥95%

Supplier: Matrix Scientific

115948-1G 115948-10G
102899-966EA 2307.69 USD
102899-966 102899-970
(3S)-1-(Oxetan-3-yl)piperidin-3-amine ≥95%
(3S)-1-(Oxetan-3-yl)piperidin-3-amine

Hazard: Irritant

Formula: C₈H₁₆N₂O
MDL Number: MFCD20233074
CAS Number: 1349699-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR