N-(Oxetan-3-yl)piperidin-4-amineoxalate ≥95%

Supplier: Matrix Scientific

115947-5G
102899-962EA 5015.38 USD
102899-962
N-(Oxetan-3-yl)piperidin-4-amineoxalate ≥95%
N-(Oxetan-3-yl)piperidin-4-amineoxalate

Hazard: Irritant

Formula: C₈H₁₆N₂O
MW: 156.23 g/mol
MDL Number: MFCD24368563
CAS Number: 1228948-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR