2-(Oxetan-3-yloxy)pyrimidin-5-ylamine ≥95%

Supplier: Matrix Scientific

115944-10G 115944-1G 115944-5G
102898-862EA 2087.69 USD
102898-862 102898-864 102898-866
2-(Oxetan-3-yloxy)pyrimidin-5-ylamine ≥95%
2-(Oxetan-3-yloxy)pyrimidin-5-ylamine

Hazard: Irritant

Formula: C₇H₉N₃O₂
MDL Number: MFCD20233082
CAS Number: 1349709-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR