2-(Oxetan-3-yloxy)pyridin-3-amine ≥95%
Supplier: Matrix Scientific
115941-10G
115941-1G
102899-946EA
10080
USD
102899-946
102899-942
2-(Oxetan-3-yloxy)pyridin-3-amine ≥95%
2-(Oxetan-3-yloxy)pyridin-3-amine
Hazard: Irritant
Formula:
C₈H₁₀N₂O₂ |
MDL Number:
MFCD20233080 CAS Number: 1349718-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...