[4-(Oxetan-3-yloxy)phenyl]methanamine ≥95%

Supplier: Matrix Scientific

115940-1G
102899-926EA 1680 USD
102899-926
[4-(Oxetan-3-yloxy)phenyl]methanamine ≥95%
[4-(Oxetan-3-yloxy)phenyl]methanamine

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD20233039
CAS Number: 1349719-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR