6-Oxa-3-azabicyclo[3.1.1]heptane tosylate ≥95%

Supplier: Matrix Scientific

115928-250MG 115928-500MG 115928-1G
102898-820EA 1238.46 USD
102898-820 102898-822 102898-824
6-Oxa-3-azabicyclo[3.1.1]heptane tosylate ≥95%
6-Oxa-3-azabicyclo[3.1.1]heptane tosylate

Hazard: Irritant

Formula: C₅H₉NO
MDL Number: MFCD27665413
CAS Number: 1339953-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR