3-(4-Nitrophenoxy)oxetane ≥95%

Supplier: Matrix Scientific

115924-10G 115924-5G
102899-010EA 6907.69 USD
102899-010 102899-884
3-(4-Nitrophenoxy)oxetane ≥95%
3-(4-Nitrophenoxy)oxetane

Hazard: Irritant

Formula: C₉H₉NO₄
MDL Number: MFCD20233112
CAS Number: 1356114-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR