4-(3-Methyloxetan-3-yl)piperidine ≥95%
Supplier: Matrix Scientific
115917-500MG
115917-250MG
115917-1G
102898-794EA
4458.46
USD
102898-794
102898-994
102898-792
4-(3-Methyloxetan-3-yl)piperidine ≥95%
4-(3-Methyloxetan-3-yl)piperidine
Hazard: Irritant
Formula:
C₉H₁₇NO |
MDL Number:
MFCD20669549 CAS Number: 873405-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...