3-(4-Methoxyphenyl)-3-oxetanamine hydrochloride ≥95%

Supplier: Matrix Scientific

115911-5G 115911-1G 115911-10G
102898-784EA 1787.69 USD
102898-784 102898-786 102898-788
3-(4-Methoxyphenyl)-3-oxetanamine hydrochloride ≥95%
3-(4-Methoxyphenyl)-3-oxetanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD19381734
CAS Number: 1332765-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR