[3-Methoxy-4-(oxetan-3-yloxy)phenyl]methanamine ≥95%
Supplier: Matrix Scientific
115910-1G
115910-500MG
115910-5G
102898-782EA
5767.69
USD
102898-782
102898-778
102898-780
[3-Methoxy-4-(oxetan-3-yloxy)phenyl]methanamine ≥95%
[3-Methoxy-4-(oxetan-3-yloxy)phenyl]methanamine
Hazard: Irritant
Formula:
C₁₁H₁₅NO₃ |
MDL Number:
MFCD20233040 CAS Number: 1349718-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...