3-Fluoro-4-(oxetan-3-yloxy)benzonitrile ≥95%
Supplier: Matrix Scientific
115893-1G
115893-10G
115893-5G
102898-652EA
1978.46
USD
102898-652
102898-654
102898-656
3-Fluoro-4-(oxetan-3-yloxy)benzonitrile ≥95%
3-Fluoro-4-(oxetan-3-yloxy)benzonitrile
Hazard: Irritant
Formula:
C₁₀H₈FNO₂ |
MDL Number:
MFCD20233108 CAS Number: 1349717-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...