N-(2-Fluorobenzyl)oxetan-3-amine ≥95%
Supplier: Matrix Scientific
115887-10G
115887-5G
102898-642EA
4290.77
USD
102898-642
102899-832
N-(2-Fluorobenzyl)oxetan-3-amine ≥95%
N-(2-Fluorobenzyl)oxetan-3-amine
Hazard: Irritant
Formula:
C₁₀H₁₂FNO |
MDL Number:
MFCD18849224 CAS Number: 1340533-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...