(4,4-Dimethyloxetan-2-yl)methylamine ≥95%

Supplier: Matrix Scientific

115881-100MG 115881-500MG
102899-828EA 1044.62 USD
102899-828 102898-764
(4,4-Dimethyloxetan-2-yl)methylamine ≥95%
(4,4-Dimethyloxetan-2-yl)methylamine

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD22628734
CAS Number: 1408075-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR