3-(4-Chlorophenyl)-3-oxetanamine hydrochloride ≥95%

Supplier: Matrix Scientific

115873-5G 115873-1G
102898-536EA 4701.54 USD
102898-536 102899-812
3-(4-Chlorophenyl)-3-oxetanamine hydrochloride ≥95%
3-(4-Chlorophenyl)-3-oxetanamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁Cl₂NO
MDL Number: MFCD17015007
CAS Number: 1245896-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR