3-(3-Chlorophenyl)-3-oxetanamine hydrochloride ≥95%
Supplier: Matrix Scientific
115872-1G
115872-500MG
115872-5G
102898-532EA
1238.46
USD
102898-532
102898-534
102899-808
3-(3-Chlorophenyl)-3-oxetanamine hydrochloride ≥95%
3-(3-Chlorophenyl)-3-oxetanamine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁Cl₂NO |
MDL Number:
MFCD19381730 CAS Number: 1332920-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...