3-(4-Bromophenyl)oxetan-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Bromophenyl)-1,3-epoxypropan-2-amine hydrochloride3-Amino-3-(4-bromophenyl)oxetane hydrochloride

115854-1G 115854-250MG 115854-500MG
102898-556EA 1867.69 USD
102898-556 102898-558 102898-560
3-(4-Bromophenyl)oxetan-3-amine hydrochloride ≥95%
3-(4-Bromophenyl)oxetan-3-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀BrNO·ClH
MW: 264.547 g/mol
MDL Number: MFCD19687804
CAS Number: 1349972-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR