2-(Bromomethyl)-3-fluoropyridine hydrochloride ≥95%

Supplier: Matrix Scientific

115851-1G 115851-500MG
102898-492EA 866.15 USD
102898-492 102898-494
2-(Bromomethyl)-3-fluoropyridine hydrochloride ≥95%
2-(Bromomethyl)-3-fluoropyridine hydrochloride

Hazard: Irritant

Formula: C₆H₆BrClFN
MDL Number: MFCD22628726
CAS Number: 122307-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR