3-Aminomethyl-3-(pyrrolidin-1-yl)oxetane ≥95%

Supplier: Matrix Scientific

Synonyms: (3-(Pyrrolidin-1-yl)oxetan-3-yl)methanamine

115830-500MG
102900-276EA 1550.77 USD
102900-276
3-Aminomethyl-3-(pyrrolidin-1-yl)oxetane ≥95%
3-Aminomethyl-3-(pyrrolidin-1-yl)oxetane

Hazard: Irritant

Formula: C₈H₁₆N₂O
MW: 156.23 g/mol
MDL Number: MFCD22628718
CAS Number: 1416323-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR