3-Aminomethyl-3-(3-bromophenyl)oxetane ≥95%

Supplier: Matrix Scientific

115828-500MG
102900-264EA 3438.46 USD
102900-264
3-Aminomethyl-3-(3-bromophenyl)oxetane ≥95%
3-Aminomethyl-3-(3-bromophenyl)oxetane

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD22546741
CAS Number: 1363380-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR