6-Hydroxymethyl-3-azabicyclo[3.1.1]heptane ≥95%
Supplier: Matrix Scientific
115779-1G
115779-500MG
102897-510EA
5144.62
USD
102897-510
102897-508
6-Hydroxymethyl-3-azabicyclo[3.1.1]heptane ≥95%
6-Hydroxymethyl-3-azabicyclo[3.1.1]heptane
Hazard: Irritant
Formula:
C₇H₁₃NO MW: 127.19 g/mol |
MDL Number:
MFCD22573390 CAS Number: 1389264-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...