6-Bromo-2,4-dichlorofuro[3,2-d]pyrimidine ≥95%
Supplier: Matrix Scientific
115759-500MG
115759-1G
115759-250MG
102897-426EA
1593.85
USD
102897-426
102897-428
102897-430
6-Bromo-2,4-dichlorofuro[3,2-d]pyrimidine ≥95%
6-Bromo-2,4-dichlorofuro[3,2-d]pyrimidine
Hazard: Irritant
Formula:
C₆HBrCl₂N₂O |
MDL Number:
MFCD20258032 CAS Number: 1363382-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...