1-Azido-5-iodonaphthalene ≥95%
Supplier: Matrix Scientific
115755-500MG
115755-1G
115755-250MG
102897-404EA
2016.92
USD
102897-404
102897-402
102897-406
1-Azido-5-iodonaphthalene ≥95%
1-Azido-5-iodonaphthalene
Hazard: Irritant
Formula:
C₁₀H₆IN₃ |
MDL Number:
MFCD22573372 CAS Number: 63785-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...