3-(Boc-amino)-4-trifluoromethylpyridine ≥95%

Supplier: Matrix Scientific

115750-500MG 115750-1G
102897-378EA 2172.31 USD
102897-378 102897-376
3-(Boc-amino)-4-trifluoromethylpyridine ≥95%
3-(Boc-amino)-4-trifluoromethylpyridine

Hazard: Irritant

Formula: C₁₁H₁₃F₃N₂O₂
MDL Number: MFCD22573369
CAS Number: 1392804-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR