1-Triisopropylsilany-5-(Boc-amino)-7-azaindole ≥95%

Supplier: Matrix Scientific

115734-1G
102897-272EA 2226.15 USD
102897-272
1-Triisopropylsilany-5-(Boc-amino)-7-azaindole ≥95%
1-Triisopropylsilany-5-(Boc-amino)-7-azaindole

Hazard: Irritant

Formula: C₂₁H₃₅N₃O₂Si
MDL Number: MFCD16039205
CAS Number: 651744-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR