Tetrahydro-1H-pyrrolizine-7a(5H)-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Hexahydro-1H-pyrrolizine-7a-carboxylic acid hydrochloride

115733-1G
102897-270EA 2363.08 USD
102897-270
Tetrahydro-1H-pyrrolizine-7a(5H)-carboxylic acid hydrochloride ≥95%
Tetrahydro-1H-pyrrolizine-7a(5H)-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₄ClNO₂
MW: 191.66 g/mol
MDL Number: MFCD11035722
CAS Number: 165456-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR