(R)-tert-Butyl-2-(pyrrolidin-2-yl)ethylcarbamate ≥95%

Supplier: Matrix Scientific

115729-1G
102900-224EA 1073.85 USD
102900-224
(R)-tert-Butyl-2-(pyrrolidin-2-yl)ethylcarbamate ≥95%
(R)-tert-Butyl-2-(pyrrolidin-2-yl)ethylcarbamate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD06656825
CAS Number: 720000-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR