1H-Pyrazolo[4,3-b]pyridine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

115727-1G
102899-612EA 3515.38 USD
102899-612
1H-Pyrazolo[4,3-b]pyridine-5-carboxylic acid ≥95%
1H-Pyrazolo[4,3-b]pyridine-5-carboxylic acid

Hazard: Irritant

Formula: C₇H₅N₃O₂
MDL Number: MFCD18250817
CAS Number: 1260670-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR